β-Cyclocitral ≥90%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B25700.18 B25700.09
B25700.18EA 8050 DKK
B25700.18 B25700.09
β-Cyclocitral ≥90%
β-Cyclocitral

Purity: 90+% CAS No.: 432-25-7 Molecular Formula: C10H16O Formula Weight: 152.23 Flash Point: 110°(230°F) Refractive Index: 1.4980 Boiling Point: 62-63°/3mm MDL No.: MFCD00079078

Formel: C₁₀H₁₆O
Kogepunkt: 62…63 °C (3 mmHg)
Massefylde: 0,944
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00079078
CAS-nummer: 432-25-7
EINECS: 207-081-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR