2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
N,N-dimethyl-3,5-di-tert-butyl-4-hydroxybenzylamine
D2301-25G
D2301-500G
TCIAD2301-25GEA
148
DKK
TCIAD2301-25G
TCIAD2301-500G
2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol ≥98.0% (by HPLC, titration analysis)
2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol
Formel:
C₁₇H₂₉NO Kogepunkt: 179°C |
MDL Number:
MFCD00026283 CAS-nummer: 88-27-7 EINECS: 201-816-1 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 93.0 to 96.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...