4,4'-Dihydroxydiphenylether ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4'-Oxydiphenol, 4,4-Oxydiphenol

D2121-25G D2121-500G
TCIAD2121-25GEA 485 DKK
TCIAD2121-25G TCIAD2121-500G
4,4'-Dihydroxydiphenylether ≥98.0% (by GC)
4,4'-Dihydroxydiphenylether
Formel: C₁₂H₁₀O₃
Molekylvægt: 202.2060 g/mol
MDL Number: MFCD00016463
CAS-nummer: 1965-09-9
EINECS: 217-809-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0618cb91fa0a4de483a67a79625adf851dd10e0&vid=e0618cb91fa0a4de483a67a79625adf851dd10e0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 166.0 to 170.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR