Choline iodide ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Choline Iodide
L00860.09
L00860.18
L00860.30
L00860.09EA
419
DKK
L00860.09
L00860.18
L00860.30
Choline iodide ≥98%
Choline iodide
Purity: 98%
CAS No.: 17773-10-3
Molecular Formula: HOCH2CH2N(CH3)3I
Formula Weight: 231.08
Melting Point: ca 270° dec.
Beilstein Reference: 3912691
Formel:
C₅H₁₄INO Smeltepunkt: ∼ 270 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011900 CAS-nummer: 17773-10-3 EINECS: 241-754-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...