Bis(benzonitrile)palladium(II) dichloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Bis(benzonitrile)palladium chloride, Dichlorobis(benzonitrile)palladium(II), Bis(benzonitrile)palladium(II) Dichloride
B1668-1G
B1668-5G
TCIAB1668-1GEA
1650
DKK
TCIAB1668-1G
TCIAB1668-5G
Bis(benzonitrile)palladium(II) dichloride ≥98.0% (by titrimetric analysis)
Bis(benzonitrile)palladium(II) dichloride
Formel:
C₁₄H₁₀Cl₂N₂Pd Molekylvægt: 383.57 g/mol |
MDL Number:
MFCD00013123 CAS-nummer: 14220-64-5 EINECS: 238-085-3 UN nummer: 3439 ADR: 6.1 III |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...