Life science sealing films For PCR and storage
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
701367 701376 701377
210-0005EA 1130 DKK
210-0005 732-3899 391-0703
Life science sealing films For PCR and storage
Forsegling Mikrotiterpladetætninger
This film is ideal for PCR, ELISA and sample storage. Due to its adhesive, it can be used down to –80 °C, is DMSO resistant and resistant to common solvents.
  • Temperature stability up to 120 °C
  • Tight seal for minimising evaporation
  • DMSO and solvent resistant
Two of the foils (701376 and 701377) can be removed without leaving any residue so that work can continue unhindered after transport or storage.