PerfeCTa® MultiPlex qPCR ToughMix®
Leverandør: Quantabio
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
95147-01K 95147-05K 95147-250 95148-01K 95148-05K 95148-250 95149-01K 95149-05K 95149-250
733-2322EA 21580 DKK
733-2322 733-2323 733-2324 733-2325 733-2326 733-2327 733-2328 733-2329 733-2330
PerfeCTa® MultiPlex qPCR ToughMix®
Nukleinsyrereagenser qPCR/RT-qPCR Enzymes and Kits
PerfeCTa® MultiPlex qPCR ToughMix® is a 5X concentrated, ready to use reaction cocktail for Real-Time quantitative PCR (qPCR) that overcomes many known inhibitors of PCR often present in crude samples extracted from environmental specimens, plant tissues, or animal tissues. It contains all components, except primers, probes and templates. The 5X concentrated ToughMix® allows addition of higher amounts of template and improved detection sensitivity with low concentration samples. PerfeCta® MultiPlex qPCR ToughMix® has been optimised to deliver maximum PCR efficiency, sensitivity, and specificity in reduced reaction volumes with fast cycle or conventional PCR cycling protocols.
  • Tough tested - overcomes common inhibitors, including polysaccharides, heme/haemoglobin, humic acid and melanin
  • Flexible - use fast or standard qPCR cycling conditions
  • Broad dynamic range - reliable data from your precious samples every time