HPLC columns, Hypersil™ SAS (C1)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30505-154630 30505-014001 30505-254630 30505-204630
555-5995EA 7020 DKK
555-5995 555-6010 555-5990 555-5994
HPLC columns, Hypersil™ SAS (C1)
Kromatografikolonner
Hypersil™ SAS (C1) columns have a short alkyl chain (C1), making it the least retentive of Hypersil™ alkyl bonded phases. They exhibit good stability for a short chain bonded phase. They exhibit unique selectivity for polar analytes and multi-functional compounds, and hence can be used in ion-pair separations.
  • Short alkyl chain reversed phase material
  • Least retentive Hypersil™ phase
  • Suitable for ion-pair separations