Kaliumhydroxid 25% (w/v) in aqueous solution
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Potash caustic, Caustic potash
46330.AP 46330.K2 46330.K7
46330.APEA 637 DKK
46330.AP 46330.K2 46330.K7
Kaliumhydroxid 25% (w/v) in aqueous solution
Potassium hydroxide
CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Physical Form: Liquid UN No.: 1814 MDL No.: MFCD00003553
Formel: KOH
Molekylvægt: 56.11 g/mol
Kogepunkt: 1320
Smeltepunkt: 360
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003553
CAS-nummer: 1310-58-3
EINECS: 215-181-3
UN nummer: 1814
ADR: 8 II
Merck Index: 12,07806