Methyl-2-amino-3-methylbenzoate ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M2193-25G M2193-5G
TCIAM2193-25GEA 911 DKK
TCIAM2193-25G TCIAM2193-5G
Methyl-2-amino-3-methylbenzoate ≥98.0% (by GC, titration analysis)
Methyl-2-amino-3-methylbenzoate
Formel: C₉H₁₁NO₂
Kogepunkt: 153°C
MDL Number: MFCD06200918
CAS-nummer: 22223-49-0
EINECS: 244-847-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dde6d3765a77185de78e4cca96c4a281955b7c9&vid=5dde6d3765a77185de78e4cca96c4a281955b7c9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Freezing point 25.0 to 29.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR