3-Anilinopropionitrile ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A0477-25G A0477-500G
TCIAA0477-25GEA 501 DKK
TCIAA0477-25G TCIAA0477-500G
3-Anilinopropionitrile ≥98.0% (by GC, titration analysis)
3-Anilinopropionitrile
Formel: C₉H₁₀N₂
Kogepunkt: 160°C
MDL Number: MFCD00001953
CAS-nummer: 1075-76-9
EINECS: 214-057-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7744edf33ee22e0d5164c3e72e2e2b02361feb38&vid=7744edf33ee22e0d5164c3e72e2e2b02361feb38&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 48.0 to 53.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR