4-(2-Bromoethyl)benzoic acid methyl ester ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-(2-Bromoethyl)benzoic acid
B1944-25G
B1944-5G
TCIAB1944-25GEA
4030
DKK
TCIAB1944-25G
TCIAB1944-5G
4-(2-Bromoethyl)benzoic acid methyl ester ≥98.0% (by titrimetric analysis)
4-(2-Bromoethyl)benzoic acid methyl ester
Formel:
C₉H₉BrO₂ |
MDL Number:
MFCD00016541 CAS-nummer: 52062-92-7 EINECS: 257-632-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...