4-Butylbenzamide ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A19516.03 A19516.06
A19516.03EA 308 DKK
A19516.03 A19516.06
4-Butylbenzamide ≥97%
4-Butylbenzamide

Purity: 97% CAS No.: 107377-07-1 Molecular Formula: C11H15NO Formula Weight: 177.25 Melting Point: 127-129° MDL No.: MFCD00221466

Formel: C₁₁H₁₅NO
Molekylvægt: 177.25 g/mol
Smeltepunkt: 127…129 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00221466
CAS-nummer: 107377-07-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR