3-Amino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A0914-25G
A0914-5G
TCIAA0914-25GEA
1460
DKK
TCIAA0914-25G
TCIAA0914-5G
3-Amino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)
3-Amino-2,2-dimethyl-1-propanol
Formel:
C₅H₁₃NO |
MDL Number:
MFCD00059847 CAS-nummer: 26734-09-8 EINECS: 247-948-3 UN nummer: 3263 ADR: 8,III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Solubility in Water | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...