4-Isoquinolineboronic acid pinacol ester ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline

I0739-1G I0739-5G
TCIAI0739-1GEA 567 DKK
TCIAI0739-1G TCIAI0739-5G
4-Isoquinolineboronic acid pinacol ester ≥98.0% (by GC)
4-Isoquinolineboronic acid pinacol ester
Formel: C₁₅H₁₈BNO₂
MDL Number: MFCD05663891
CAS-nummer: 685103-98-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe24443c6999ab41256bdae4b475793c770ebb02&vid=fe24443c6999ab41256bdae4b475793c770ebb02&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR