6-Bromo-2-methoxyquinoline 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H54226.03
H54226.06
H54226.03EA
1920
DKK
H54226.03
H54226.06
6-Bromo-2-methoxyquinoline 96%
6-Bromo-2-methoxyquinoline
Purity: 96%
CAS No.: 99455-05-7
Molecular Formula: C10H8BrNO
Formula Weight: 238.08
Melting Point: 87-89°
MDL No.: MFCD11847805
Formel:
C₁₀H₈BrNO Molekylvægt: 238.08 g/mol Smeltepunkt: 87…89 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11847805 CAS-nummer: 99455-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...