Industrial gloves, GemoMech® 665
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
665.6 665.7 665.8 665.9 665.10 665.11
KCHL665.6EA 85.3 DKK
KCHL665.6 KCHL665.7 KCHL665.8 KCHL665.9 KCHL665.10 KCHL665.11
Industrial gloves, GemoMech® 665
Handsker
These polyamide gloves feature a nitrile foam coating on palm and fingers and additional nitrile dots on the palm.
  • Good grip when handling greasy and oily parts
  • Flexible, very durable
  • Excellent mechanical resistance
Perfect for assembly works, automotive and supply industries, metal and plastics processing, storage and commission works and maintenance and repair work.
Thickness: 1,30 ± 0,20 mm
Colour: Black/grey
EN 388 (4121); Cat. II
Certificeringer: EN 388 (3121); Cat. II