Cut resistant gloves, Camapur® Cut 620
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620.9 620.10 620.11 620.6 620.7 620.8
KCHL620.9EA 33350 DKK
KCHL620.9 KCHL620.10 KCHL620.11 KCHL620.6 KCHL620.7 KCHL620.8
Cut resistant gloves, Camapur® Cut 620
Handsker
These seamless HPPE lined gloves feature a knit wrist and a protective PU coating on the palm.
  • PU coating ensures good grip - even when handling wet, slippery or slightly oily parts
  • Elasticated knit wrist for wearing comfort
  • Moisture-repellent, high mechanical resistance
  • Free from the allergenic substances thiurames and dithiocarbamates
Ideal for assembly works with medium cut risks, metal sheet processing and the processing of synthetic material profiles.
Thickness: 1,2 ± 0,1 mm
Colour: White/grey
EN 388 (4342), Cat. II
Certificeringer: More Product Information
Bestil nu
Specifikationer
Model Cut 620
Ambidextrous/Hånd-specifik Hånd-specifik
Environmentally Preferable
Engangs/Genbrugelig Genanvendelig
Liner material HPPE
Finish Coating on palm and fingers
Cuff style Strikket
EN norm EN 388 (4342)