Bottles for dispensers
Leverandør: VITLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1671505 1671510 1671506 1671515 1671520 1671500
VITL1671505EA 84 DKK
VITL1671505 215-2966 VITL1671506 VITL1671515 215-2964 215-2965
Bottles for dispensers
Flasker Dråbeflasker
Soda lime glass, brown, coated.
  • Plastic coating significantly reduces hazardous glass splintering during breakage
  • Maximum working temperature for coated bottles is 80 °C
  • To preserve the coating, do not clean at temperatures exceeding 60 ºC
Threaded brown glass (soda lime glass) bottles with an ethylene acrylate coating for increased safety, and a screw cap.