Prøvebeholdere med skruelåg, Bijou, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
327145 327147 327149
216-0979EA 1150 DKK
216-0979 216-0980 216-0981
Prøvebeholdere med skruelåg, Bijou, WHEATON®
Prøvecontainere Allround kasser
PS clear, with PE screw cap.
  • Manufactured from FDA approved virgin PS
  • Supplied sterile and for in vitro use only
  • PS offers good thermal and chemical resistance
  • All materials are non-cytotoxic
Straight-walled vessels ideal for sample collection, storage and transport. Supplied with leak proof, flow seal closure. Available pre-filled with boric acid for urine samples.
Certificeringer: CE marked in accordance with European Directive 98/79/EC
All containers are leak tested to British Standard BS5213:1975(1989)
Emballering: Packed in inner bags of 50.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904