Cleanroom T-shirts, 7R015
Leverandør: FRISTADS KANSAS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1006415202XL 1006415203XL 100641520L 100641520M 100641520S 100641520XL 100641520XS
KWIN1006415202XLEA 327 DKK
KWIN1006415202XL KWIN1006415203XL KWIN100641520L KWIN100641520M KWIN100641520S KWIN100641520XL KWIN100641520XS
Cleanroom T-shirts, 7R015
Arbejdstøj
Shirts made of XA80 fabric consisting of 100% filament polyester with a moisture transport system. The structure is reminiscent of fine silk.
  • Short-sleeved
  • Crew neck
  • Moisture-wicking
  • Suitable as underwear
Tests have shown that the material also functions as a pre-filter. Complete underwear worn in combination with cleanroom workwear boosts particle filtration by up to 50%.
Weight: 99 g/m²