Pipette tips, ART™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3550 3520 3500 3552-RT 3502-HR 3101-HR 3101-RT 3102-HR 3102-RT 3501-HR 3501-RT 3502-RT 3521-HR 3521-RT 3522-HR 3522-RT 3551-HR 3551-RT 3552-HR 3771-HR 3771-RT 3772-HR 3772-RT
612-9748EA 134 DKK
612-9748 612-9749 612-9751 613-2496 613-2464 613-2448 613-2449 613-2452 613-2453 613-2460 613-2461 613-2465 613-2472 613-2473 613-2476 613-2477 613-2491 613-2492 613-2495 613-2499 613-2774 613-2779 613-2780
Pipette tips, ART™
Pipettespidser
These non filtered tips are designed to fit virtually any micropipette. They are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.
  • Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
  • Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks ensure safe and secure storage
  • Space saving reload system is environmentally friendly, traceable and easy to use
Emballering: Tips are available in bulk packaging, in racks or in trays for rack reloading. Reload tips require an empty rack.