Filter plates, 96-well, for protein purification, AcroPrep™ Advance
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8129 8072 8071 8039 8130 8172 8171 8022
738-0117EA 1680 DKK
738-0117 738-0114 738-0113 518-0024 738-0118 738-0128 738-0127 738-0197
Filter plates, 96-well, for protein purification, AcroPrep™ Advance
Mikroplader Filterplader
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications; gross fractionation and lysate clarification and general filtration.
  • Supor® membranen giver optimal støtte til at bevare kromotografi resinen samtidig med at den giver mulighed