Filter plates, 96-well, for ultrafiltration, AcroPrep™ Advance
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8034 8036 8035 8033 8166 8165 8164 8163
518-0032EA 5330 DKK
518-0032 518-0029 518-0028 518-0027 738-0126 738-0125 738-0124 738-0123
Filter plates, 96-well, for ultrafiltration, AcroPrep™ Advance
Mikroplader Filterplader
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications, gross fractionation and lysate clarification and general filtration.
  • Høj genindvinding af biomolekyler - Omega® membran, der typisk resulterer i ≥90 % genindvinding af mål-biomolekyler
  • Brøndgeometrien resulterer i hurtigere og mere ensartede filtreringsrater på tværs af pladen med formindsket retentionsvolumen
  • Lav uspecifik binding - inerte plade- og membranegenskaber forhindrer, at målmolekylerne binder til pladen
Applications for AcroPrep™ Advance 96-well filter plates for ultrafiltration include size exclusion, PCR clean-up, nucleic acid purification, and protein separation.