Filter plates, 96-well, for lysate clearance, AcroPrep™ Advance
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8075 8040 8275 8175
518-0031EA 3170 DKK
518-0031 518-0030 738-0133 738-0129
Filter plates, 96-well, for lysate clearance, AcroPrep™ Advance
Mikroplader Filterplader
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications; gross fractionation and lysate clarification; and general filtration.
  • Integrated prefilter yields consistent filtration of samples with high levels of gross particulate
  • Well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume
  • Output tip geometry provides direct flow of samples into receiver plate without concerns about cross-contamination
  • Biologically inert materials allows clarification of most types of lysates without loss of target molecules
Applications for AcroPrep™ Advance 96-well filter plates for lysate clearance include removal of bacterial debris prior to plasmid purification, removal of bacterial and cellular debris prior to protein purification, and clearance of gross particulates.