Filter plates, 96-well, for DNA purification, AcroPrep™ Advance
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8132 8032
738-0120EA 1520 DKK
738-0120 738-0108
Filter plates, 96-well, for DNA purification, AcroPrep™ Advance
Mikroplader Filterplader
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications; gross fractionation and lysate clarification, and general filtration.
  • Høj bindingskapacitet - optimeret for maks. binding og DNA-udbytte fra en lang række prøvetyper
  • Brøndenes