Pumper til omhældning, eller fodbetjent, OTAL®
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5000-1500 5000-2200 5000-4016 5000-5018 5000-1800 5000-3012 5000-6000
223-0042EA 1910 DKK
223-0042 223-1122 223-0004 223-0005 223-1123 223-1124 223-0117
Pumper til omhældning, eller fodbetjent, OTAL®
Pumper Tromlepumper
The telescopic immersion tube adapts itself to the container height on insertion of the foot pump. This ensures good removal of residual liquid and means that the foot pump can be used with different containers.
  • Simple handling: Both hands remain free for filling
  • Very good pump capacity due to large volume foot bellows
  • With universal bungs made of soft PVC, for container openings of 30 to 70 mm Ø
OTAL® foot pump made of PP
OTAL® foot pumps with delivery tubes made of PP, resistant to weak acids, alkalis and detergents.
OTAL® foot pump made of PVDF
OTAL® foot pump with delivery tubes made of PVDF, particularly for acids and alkalis, not for heavily oxidising acids and alkalis. Resistant to chlorine bleach. With its special material properties, PVDF has an even better chemical resistance than PP.
OTAL® foot pump made of stainless steel
OTAL® foot pump made of stainless steel AISI 304 (1,4301), with delivery tubes made of stainless steel for liquids that are not easily combustible in Hazard Category AIII (VbF).
OTAL® foot pump with hose and stopcock
OTAL® foot pump with hose and tap with delivery tubes made of PP and a 1,5 m discharge hose made of PVC with stopcock made of PP/PE Ø 10 mm.
OTAL® battery-pump
OTAL® pump for safe filling of car batteries with acid or distilled water, with hose shutoff valve and dosing tip for precise filling.