Safety spectacles, KLEENGUARD* V20
Leverandør: KIMBERLY CLARK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25654 25656
KIMB25654EA 47.8 DKK
KIMB25654 KIMB25656
Safety spectacles, KLEENGUARD* V20
Briller
A stylish new standard in eye protection that improves personal comfort, safety and worker performance. KLEENGUARD* V20 Comfort Eye Protection features distortion-free lenses with proven impact protection, added comfort and a broad range of styles.
  • 99,9% UVA/UVB protection
  • Wrap-around lens for better eye coverage
  • Comfortable, padded temples
Certificeringer: EN 166; 1F (111-9010: 1FT); ANSI Z87.1, CSA Z94.3