Fleece lining for jackets, flame-retardant, Molenbaix 7761
Leverandør: SIOEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7761/B75/1XL 7761/B75/2XL 7761/B75/3XL 7761/B75/L 7761/B75/M 7761/B75/S
SIOE7761/B75/1XLEA 338 DKK
SIOE7761/B75/1XL SIOE7761/B75/2XL SIOE7761/B75/3XL SIOE7761/B75/L SIOE7761/B75/M SIOE7761/B75/S
Fleece lining for jackets, flame-retardant, Molenbaix 7761
Arbejdstøj
Detachable lining without sleeves, made of double-sided, flame-retardant polyester with anti-pilling treatment. Can be zipped into different jackets.
  • Good cold protection, breathable
  • Round neck, zip closure
  • One patched chest pocket, one inside pocket
  • Without sleeves
  • Can not be worn separately
Weight: 275 g/m²
Certificeringer: EN ISO 14116 (Index 1), EN 14058 (1XXXX)