Paddle tester, total aerobic bacteria/yeast and mould
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2610810 2610910 2619510
HACH2610810EA 366 DKK
HACH2610810 HACH2610910 HACH2619510
Paddle tester, total aerobic bacteria/yeast and mould
Mikrobiologiske testsystemer Test for mikroorganismer
Hach Paddle testers allow fast, easy screening of water samples, solid surfaces, and non viscous liquids where unsanitary conditions may exist. The paddle is a double-sided slide attached to the vial cap. Each side of the slide is used to perform a separate test. The slide has a moulded-in grid for easy colony counting without opening the optically clear, leakproof vial. Paddle tester, total aerobic bacteria/yeast and mould (tryptone glucose extract agar/sabouraud dextrose agar).
  • Save time and simplify lab and field testing with Hach prepared P/A media
  • Report total coliform values with EPA accepted Presence-Absence bottles and media
  • Simplify routine testing with self-contained, easy-to-use paddle testers and BART™ products
Certificeringer: UL Listed.