Safety storage cabinets, type 90, Q-CLASSIC-90
Leverandør: asecos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30001040 30001041 30001046 30001047 30001048 30001049 30001050 30041040 30041041 30041046 30041047 30041048 30041049 30041050 30072040 30072041 30072046 30072047 30072048 30072049 30072050 30073040 30073041 30073046 30073047 30073048 30073049 30073050 37586040 3000104033640 37587040
135-2181EA 28100 DKK
135-2181 135-1742 135-2182 135-2183 135-2184 135-2185 135-2186 135-1749 135-2270 135-2366 135-2367 135-2368 135-2369 135-2370 135-2242 135-1753 135-2388 135-2389 135-2390 135-2391 135-2392 135-2393 135-2394 135-2395 135-2396 135-2397 135-2398 135-2399 MSGA37586040 MSGA3000104033640 MSGA37587040
Safety storage cabinets, type 90, Q-CLASSIC-90
Møbler Opbevaringskabinetter
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe, durable, robust and convenient with transport base and bicoloured cabinet design.
  • Robust construction and longevity – triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch and impact resistant surface, easy to clean
  • Easy handling with comfort – doors stop in any position, self-closing in the event of fire
  • No unauthorised use – doors lockable with cylinder locking (integration in a locking system possible) and locking state indicator (red/green)
  • Easy transport – integrated transport base with removable, optional base cover
  • Easy alignment – adjusting aids to compensate for uneven floor
  • Quickly put into operation – integrated exhaust outlet ready for connection (NW 75 mm) to a technical exhaust system
The 2-colour look with a choice of 7 door colours, at no extra cost, offers maximum individuality and a modern appearance.
Furnace tested (type test) in accordance with EN 14470-1
Fire resistance 90 minutes
GS approval, CE conformity
EN 14727 conformity (laboratory furniture)
Certificeringer: Furnace tested (type test) in accordance with EN 14470-1
Fire resistance 90 minutes
GS approval, CE conformity
EN 14727 conformity (laboratory furniture)
Bestillingsoplysninger: Seven door colours are available. Supplied with or without interior equipment packages. Interior equipment packages need to be ordered separately. Accessory interior equipment package MSGA34617 (135-2878) can be only ordered for the safety cabinet MSGA37586040. Please check the website or contact your local Avantor office for further information.
Leveringsinformation: Delivery prices available upon request.