VWR® Graduated Pipettes, Type 3, Class AS
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
612-4767EA 160 DKK
612-4767 612-4768 612-4769 612-4770 612-4771 612-4772 612-4773 612-4774
VWR® Graduated Pipettes, Type 3, Class AS
Pipetter Pipetter med inddeling
Borosilicate 9.1 glass.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in amber stain
  • Cotton plug ending (cotton plugs not included)
Certificeringer: DIN 12696, ISO 835
Bestillingsoplysninger: Includes a lot certificate.