High visibility traffic jacket, S460
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S460YERXXXL S460YERXXL S460YERXL S460YERS S460YERM S460YERL S460YER5XL S460YER4XL S460YER6XL S460YER7XL S460YERXS S460ORR4XL S460ORR5XL S460ORRL S460ORRM S460ORRS S460ORRXL S460ORRXXL S460ORRXXXL
PRTWS460YERXXXLEA 347 DKK
PRTWS460YERXXXL PRTWS460YERXXL PRTWS460YERXL PRTWS460YERS PRTWS460YERM PRTWS460YERL PRTWS460YER5XL PRTWS460YER4XL PRTWS460YER6XL PRTWS460YER7XL PRTWS460YERXS PRTWS460ORR4XL PRTWS460ORR5XL PRTWS460ORRL PRTWS460ORRM PRTWS460ORRS PRTWS460ORRXL PRTWS460ORRXXL PRTWS460ORRXXXL
High visibility traffic jacket, S460
Arbejdstøj
This fully certified waterproof jacket is a popular option across many industries. Made of PU coated polyester, 300D Oxford Weave, with a stain resistant finish. With nylon lining and polyester filling for additional warmth.
  • Taped waterproof seams
  • Two-way zip front with studded storm flap
  • Drawstring hood and hem, knitted storm cuffs
  • Two hip pockets with Velcro fastened flaps
  • Inside pocket, mobile phone pocket
Weight (outer fabric): 190 g/m²
Weight (nylon lining): 60 g/m²
Weight (polyester filling): 170 g/m²
Certificeringer: EN ISO 20471 (Class 3), EN 343 (3-1)