ESD safety shoes, clogs, Compositelite™ ESD FC03
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FC03BKR38 FC03BKR39 FC03BKR40 FC03BKR41 FC03WHR45 FC03WHR47 FC03WHR36 FC03WHR39 FC03WHR40 FC03WHR46 FC03WHR41 FC03WHR37 FC03WHR38 FC03WHR42 FC03WHR43 FC03WHR44 FC03BKR36 FC03BKR44 FC03BKR45 FC03BKR46 FC03BKR47 FC03BKR37 FC03BKR42 FC03BKR43
PRTWFC03BKR38EA 464 DKK
PRTWFC03BKR38 PRTWFC03BKR39 PRTWFC03BKR40 PRTWFC03BKR41 PRTWFC03WHR45 PRTWFC03WHR47 PRTWFC03WHR36 PRTWFC03WHR39 PRTWFC03WHR40 PRTWFC03WHR46 PRTWFC03WHR41 PRTWFC03WHR37 PRTWFC03WHR38 PRTWFC03WHR42 PRTWFC03WHR43 PRTWFC03WHR44 PRTWFC03BKR36 PRTWFC03BKR44 PRTWFC03BKR45 PRTWFC03BKR46 PRTWFC03BKR47 PRTWFC03BKR37 PRTWFC03BKR42 PRTWFC03BKR43
ESD safety shoes, clogs, Compositelite™ ESD FC03
Sko og støvler
Metal-free, lightweight, ESD safety clog with perforated upper for added breathability, a foldable and removable elasticated heel strap plus an oil resistant, dual-density PU sole.
  • Shock absorbent heel
  • Composite toecap (200 J)
Suitable for use in ESD environments.
Colour: Black or white
These shoes do not utilise animal product or biproducts.
Certificeringer: EN 61340 (ESD); EN ISO 20345:2001; SB, AE, SRC.