Safety shoes, lace-up, Compositelite™ WORK FC14
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FC14BKR46 FC14BKR47 FC14BKR45 FC14BKR44 FC14BKR43 FC14BKR42 FC14BKR41 FC14BKR40 FC14BKR39 FC14BKR38 FC14BKR37
PRTWFC14BKR46EA 358 DKK
PRTWFC14BKR46 PRTWFC14BKR47 PRTWFC14BKR45 PRTWFC14BKR44 PRTWFC14BKR43 PRTWFC14BKR42 PRTWFC14BKR41 PRTWFC14BKR40 PRTWFC14BKR39 PRTWFC14BKR38 PRTWFC14BKR37
Safety shoes, lace-up, Compositelite™ WORK FC14
Sko og støvler
Metal-free, anti-static, lightweight safety shoe made from split leather upper plus an oil resistant, dual-density sole.
  • Shock-absorbent heel
  • Composite toecap (200 J)
  • Puncture resistant composite midsole
Colour: Black.
Certificeringer: EN 20345:2011 + A1:2007; S1P, SRC.