Safety boots, wellingtons, Portwest WORK Classic FW 94
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FW94BKR37 FW94BKR38 FW94BKR39 FW94BKR40 FW94BKR41 FW94BKR42 FW94BKR43 FW94BKR44 FW94BKR45 FW94BKR46 FW94BKR47
PRTWFW94BKR37EA 231 DKK
PRTWFW94BKR37 PRTWFW94BKR38 PRTWFW94BKR39 PRTWFW94BKR40 PRTWFW94BKR41 PRTWFW94BKR42 PRTWFW94BKR43 PRTWFW94BKR44 PRTWFW94BKR45 PRTWFW94BKR46 PRTWFW94BKR47
Safety boots, wellingtons, Portwest WORK Classic FW 94
Sko og støvler
Anti-static safety wellingtons manufactured from PVC/nitrile (upper and sole) to give a seamless, waterproof construction.
  • Washable nylon lining
  • Kick-off spurs
  • Protective steel toecap (200 J)
Colour: Black
Certificeringer: EN ISO 20345:2011; S4, SRC.