Safety ankle boots, lace-up, Steelite™ WORK Protector, FW10
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FW10BKR43 FW10BKR42 FW10BKR41 FW10BKR40 FW10BKR39 FW10BKR38 FW10BKR37 FW10BKR36 FW10BKR46 FW10BKR44 FW10BKR45 FW10BKR47
PRTWFW10BKR43EA 233 DKK
PRTWFW10BKR43 PRTWFW10BKR42 PRTWFW10BKR41 PRTWFW10BKR40 PRTWFW10BKR39 PRTWFW10BKR38 PRTWFW10BKR37 PRTWFW10BKR36 PRTWFW10BKR46 PRTWFW10BKR44 PRTWFW10BKR45 PRTWFW10BKR47
Safety ankle boots, lace-up, Steelite™ WORK Protector, FW10
Sko og støvler
Comfortable and sturdy anti-static safety ankle boot made from split buffalo leather plus a PU/PU dual-density sole, resistant to oil and heat (200 °C).
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black.
Certificeringer: EN ISO 20345:2011; S1P, SRC, HRO.