Flame-retardant bomber jacket with detachable sleeves, high visibility, Oregon 1874
Leverandør: SIOEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1874/278/2XL 1874/278/1XL 1874/278/3XL 1874/278/L 1874/278/M 1874/278/S
SIOE1874/278/2XLEA 1300 DKK
SIOE1874/278/2XL SIOE1874/278/1XL SIOE1874/278/3XL SIOE1874/278/L SIOE1874/278/M SIOE1874/278/S
Flame-retardant bomber jacket with detachable sleeves, high visibility, Oregon 1874
Arbejdstøj
Anti-static multi-norm bomber jacket made of Sio-Start FR AST fabric: 100% polyester fabric with 100% FR PU coating+AST. Its fixed FR cotton lining provides a lot of warmth and comfort.
  • Highly breathable, water- and windproof, high tear resistance
  • Straight collar
  • Hidden zip front closure, zipper in back lining for placing logos
  • Two patched pockets with flap, one inside pocket (in lining), loop for gas detector
  • Elasticated cuffs, drawstring in hem
  • Detachable sleeves and taped seams
  • Flame retardant reflective tape (50 mm)
The fabric is water repellent on the outside and features a moisture attracting coating on the inside.
Oregon can also be turned into a body warmer as the sleeves are detachable. Thanks to Sioens I.LS. system it can be combined with different parkas.
Back length: 75 cm
Weight: 250 g/m²
Certificeringer: EN 13034 (Type PB/6), EN 531 (A/B2/C1; with sleeves), EN 14116 (inside index 3; outside index 1), EN 1149-5, EN 471 (3-2 with sleeves, 2-2 without sleeves), EN 14058 (23XXX)