Headtops for respiratory system Versaflo™, M-400 series
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M407 M406
111-2209EA 7510 DKK
111-2209 111-2208
Headtops for respiratory system Versaflo™, M-400 series
Åndedrætsværn Åndedrætsbeskyttelsessystemer
Versaflo™ M-400 Series helmets with shrouds offer protection from a range of respiratory, head, eye and face hazards.
  • Lightweight, compact and well-balanced
  • Integrated with additional neck and shoulder coverage
  • Coated lenses provide added chemical and scratch resistance
  • Spare parts and accessories are quick and easy to replace
  • M-407: Features a shroud in flame resistant Nomex® for applications with hot particles
Certificeringer: EN 12941 TH3, EN 14594 3B, EN 166 1:B:3, EN 397