Safety shoes, clogs, HACCP, Anatom, 2120/2125
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2120/36 2120/37 2120/38 2120/39 2120/40 2120/41 2120/42 2120/43 2120/44 2120/45 2120/46 2120/47 2120/48 2125/36 2125/37 2125/38 2125/39 2125/40 2125/41 2125/42 2125/43 2125/44 2125/45 2125/46 2125/47 2125/48 2120/49 2125/49 2125/50 2125/51 2125/52 2120/50 2120/52 2120/51
114-1137EA 635 DKK
114-1137 114-1138 114-1139 114-1140 114-1141 114-1142 114-1143 114-1144 114-1145 114-1146 114-1147 114-1148 114-1149 114-7425 114-7426 114-7427 114-7428 114-7429 114-7430 114-7431 114-7432 114-7433 114-7434 114-7435 114-7436 114-7437 114-9000 114-7438 114-7540 114-7541 114-7542 114-7538 114-7424 114-7539
Safety shoes, clogs, HACCP, Anatom, 2120/2125
Sko og støvler
Anti-static safety clogs made from microfibre with a non foldable, adjustable heel strap and a fuel resistant PU outsole with high slip resistance and a self-cleaning profile.
  • Perspiration-absorbent special lining with silver fibre
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock absorbent PU midsole
  • Sanitized® treatment reduces odour and bacterial growth
  • Protective steel toecap (200 J)
  • Washable at 30 °C
Suitable for use in kitchens.
Colour: White or black
Certificeringer: DGUV 112-191; HACCP; EN ISO 20345:2011; SB.