ESD safety shoes, lace-up, Anatom, 1122
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1122/36 1122/37 1122/38 1122/39 1122/40 1122/41 1122/42 1122/43 1122/44 1122/45 1122/46 1122/47 1122/48 1122/49 1122/50
ABEB1122/36EA 718 DKK
ABEB1122/36 ABEB1122/37 ABEB1122/38 ABEB1122/39 ABEB1122/40 ABEB1122/41 ABEB1122/42 ABEB1122/43 ABEB1122/44 ABEB1122/45 ABEB1122/46 ABEB1122/47 ABEB1122/48 ABEB1122/49 ABEB1122/50
ESD safety shoes, lace-up, Anatom, 1122
Sko og støvler
ESD shoe made from velour with breathable textile inlays, and fuel resistant PU outsole, with high slip resistance and a self-cleaning profile.
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Asymmetric heel and Flexzone for a natural bedding and foot movement
  • TPU torsion waist, arch support and high stability
  • Shock-absorbent PU midsole
  • Protective steel toecap (200 J)
Colour: Black/green
Certificeringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S1, SRC.