m-FC broth
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2373220 2373250
HACH2373220EA 386 DKK
HACH2373220 HACH2373250
m-FC broth
Mikrobiologiske medier Klargjort medium
Selectivity: Faecal coliform bacteria.
  • Save valuable time and money using this prepared media for membrane filtration methods