Turbidity standards, Stablcal®
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2660149 2659749 2659753 2659849 2659842 2659949 2660449 2660549 2660649 2698042 2697942 2697953 2659853 2659742 2660142 2660242 2660542 2660153 2723353 2660442 2660642 2659942 2660249 2723342 2896742 2896842 2896942 2897042 2746353 2805342
708-0076EA 1810 DKK
708-0076 708-0083 HACH2659753 708-0080 708-0081 708-0078 708-0071 708-0069 708-0067 708-0065 708-0066 708-0001 HACH2659853 708-0084 708-0077 708-0074 708-0070 HACH2660153 708-0003 708-0072 708-0068 708-0086 708-0073 708-0064 708-0062 708-0061 708-0060 708-0059 HACH2746353 708-0085
Turbidity standards, Stablcal®
Standarder Turbiditets standarder
Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazine solutions
  • Based on formazine primary standards
  • Enhanced stability saves time and labour
  • Confidence in results