Industrial gloves, Sahara® 100
Leverandør: KCL by Honeywell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100.10 100.7 100.8 100.9
112-2973EA 101 DKK
112-2973 KCHL100.7 112-2974 112-2975
Industrial gloves, Sahara® 100
Handsker
These lightweight, flexible cotton liners with knit wrist feature a nitrile coating on palm and fingers. They provide good resistance to mechanical impacts.
  • Moisture-repellent
  • Free from silicones
  • Good discharge capacity
  • Flexibility of the raw materials and thickness of the coating ensure good sensitivity
Suitable for woodworks and gardening, metalworking and mechanical engineering, construction industry applications, assembly and transportation work and metal and plastics processing.
Thickness: 0,8 ± 0,05 mm
Colour: White/yellow
EN 388 (3111), Cat. II; Öko-Tex standard 100 Food contact approved
Certificeringer: EN 388:2016 (3111X), Cat. II; Öko-Tex standard 100
Food contact approved