Occupational shoes, clogs, rubber, 9100/9150
Leverandør: Abeba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9100/36 9100/37 9100/38 9100/39 9100/40 9100/41 9100/42 9100/43 9100/44 9100/45 9100/47 9100/46 9150/36 9150/37 9150/38 9150/39 9150/40 9150/41 9150/42 9150/43 9150/44 9150/45 9150/46 9150/47
114-2655EA 419 DKK
114-2655 114-2656 114-2657 114-2658 114-2659 114-2660 114-2661 114-2662 114-2663 114-2664 114-1198 114-1197 ABEB9150/36 ABEB9150/37 ABEB9150/38 ABEB9150/39 ABEB9150/40 ABEB9150/41 ABEB9150/42 ABEB9150/43 ABEB9150/44 ABEB9150/45 ABEB9150/46 ABEB9150/47
Occupational shoes, clogs, rubber, 9100/9150
Sko og støvler
Anti-static clogs made from smooth leather upper, a non foldable, adjustable heel strap and a rubber sole with cross/circle profile resistant to acids, bases (cleaning agents) and fuels.
  • Leather lining
  • Sanitized® treatment reduces odour and bacterial growth
  • PU midsole
  • Slip resistant special rubber sole
  • Adjustable instep area
Ideal for use in kitchens.
Colour: White or blue
Certificeringer: EN ISO 20347:2012; OB, SRC.