Hot air sterilisers, small
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E996.1 E997.1
ROTHE996.1EA 3304.14 DKK
ROTHE996.1 ROTHE997.1
Hot air sterilisers, small
Sterilisatorer Varmluft sterilisator
For dry- sterilising of instruments.
  • Glass balls are included
  • Warm-up period 15 minutes
Sterilisation time: 10 to 30 seconds. Units are fitted with glass balls and can reach a temperature of 240 °C.
Bestil nu
Specifikationer
Environmentally Preferable
Effekt 70 W
W×D×H 120×140×140 mm