Cut resistant gloves, ActivArmr® 70-765 / 70-766
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
70-765/10 70-765/11 70-765/8 70-765/9 70-766/9 70-766/7
ANSE70-765/10EA 289 DKK
ANSE70-765/10 ANSE70-765/11 ANSE70-765/8 ANSE70-765/9 ANSE70-766/9 ANSE70-766/7
Cut resistant gloves, ActivArmr® 70-765 / 70-766
Handsker
These gloves knitted from nylon, acrylic and Kevlar® fibres feature a premium leather coating on the palm.
  • Reinforced thumb and seams relocated to the backs of the fingers: Reduce exposure to cuts and offer greater protection and longer life
  • Gloves guard against minor injuries and punctures
  • Superior grip even in oily environments thanks to the premium leather coating
  • 70-766 version has a longer cuff for additional protection
  • Plaited inside for comfort with coloured polyester/cotton
Ideal for press and stamping applications, handling heavy and oily metal pieces and for handling sharp or irregular-edged metal parts.
Certificeringer: EN 388:2016 (4X43D), EN 407 (X1XXXX); Cat. III