Cut resistant gloves, HyFlex® 70-750
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
70-750/10 70-750/7 70-750/9 70-750/8
112-3726EA 223 DKK
112-3726 112-3723 112-3725 112-3724
Cut resistant gloves, HyFlex® 70-750
Handsker
These premium gloves use innovative technology for improved cut resistance, excellent flexibility and a higher level of comfort. Made from a combination of Kevlar®, Lycra® and stainless steel.
  • Provide protection from heat and light molten splashes
  • Intercept® cut resistance technology: Gloves are made from revolutionary engineered yarns with the highest cut-protection-to-weight ratio available
  • Excellent flexibility and a high level of comfort
Reserved exclusively for the premium gloves, Ansell’s cut resistance technology uses revolutionary engineeredMore Product Information
Bestil nu
Specifikationer
Model 70-750
Ambidextrous/Hånd-specifik Hånd-specifik
Environmentally Preferable
Engangs/Genbrugelig Genanvendelig
Liner material Rustfrit stål/Lycra ®/Kevlar ®
Måler 13
Cuff style Strikket
EN norm EN 388:2016 (2X4XD), EN 407 (X1XXXX)