Cut resistant gloves, HyFlex® 11-435
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-435/9 11-435/6 11-435/7 11-435/8
112-4359EA 0 DKK
112-4359 112-4356 112-4357 112-4358
Cut resistant gloves, HyFlex® 11-435
Handsker
This knitted liner made of Dyneema®/glass fibre/Lycra® and nylon features a water-based PU coating on the palm.
  • Anti-static, breathable
  • Extreme resistance to cuts and burrs
  • Seamless plaited glove for very good fit, dexterity and flexibility
  • Water-based PU enhances the dexterity and comfort of the glove, as the PU doesn't penetrate inside it
The glove is ideal for the handling of glass and metal sheets, sharp-edged objects, pre- assembling, cutting of small dry or lightly oiled parts and maintenance tasks.
Certificeringer: EN 388:2016 (4X42C); Cat. II