Cut resistant gloves, HyFlex® 11-518
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-518/10 11-518/11 11-518/6 11-518/7 11-518/8 11-518/9
112-2729EA 1620 DKK
112-2729 112-2730 112-2725 112-2726 112-2727 112-2728
Cut resistant gloves, HyFlex® 11-518
Handsker
This knitted spandex/nylon/Dyneema® glove with thin PU palm coating provides a high level of dexterity and tactility for applications with cut risks.
  • Ultra-light fabric and tailored fit
  • Knit wrist cuff style
  • Soft seamless liner allows the hands to breath and offers enhanced flexibility and fingertip sensitivity
Ideal for final assembly applications, the handling of small or ultra-fine sharp parts, machining, packaging, stamping and wire assembly.
Certificeringer: EN 388:2016 (3X31B); Cat. II